Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(9-Carbazolyl)benzeneboronic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
864377-33-3
C18H14BNO2
287.125
MFCD18072506
IDQUIFLAFFZYEX-UHFFFAOYSA-N
3-9h-carbazol-9-yl phenylboronic acid,3-9h-carbazol-9-yl phenyl boronic acid,boronic acid, 3-9h-carbazol-9-yl phenyl,3-9h-carbazol-9-yl phenyl boronic acid,3-9-carbazolyl benzeneboronic acid,3-carbazol-9-yl phenylboronic acid,3-9h-carbazol-9-yl phenyl boronic acid,3-9h-9-carbozale phenylboronic acid,3-9h-9-carbazole phenylboronic acid,3-9h-carbazol-9-yl benzeneboronic acid
56965778
(3-carbazol-9-ylphenyl)boronic acid
B(C1=CC(=CC=C1)N2C3=CC=CC=C3C4=CC=CC=C42)(O)O
864377-33-3
C18H14BNO2
287.125
MFCD18072506
IDQUIFLAFFZYEX-UHFFFAOYSA-N
3-9h-carbazol-9-yl phenylboronic acid,3-9h-carbazol-9-yl phenyl boronic acid,boronic acid, 3-9h-carbazol-9-yl phenyl,3-9h-carbazol-9-yl phenyl boronic acid,3-9-carbazolyl benzeneboronic acid,3-carbazol-9-yl phenylboronic acid,3-9h-carbazol-9-yl phenyl boronic acid,3-9h-9-carbozale phenylboronic acid,3-9h-9-carbazole phenylboronic acid,3-9h-carbazol-9-yl benzeneboronic acid
56965778
(3-carbazol-9-ylphenyl)boronic acid
B(C1=CC(=CC=C1)N2C3=CC=CC=C3C4=CC=CC=C42)(O)O
Quantity 5g
Color White
Beilstein 15530380
Formula Weight 287.13
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 3-(9-Carbazolyl)benzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C