Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetoxy-2-methylbenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
168899-58-9
C10H10O4
194.19
LHVJUPHCLWIPLZ-UHFFFAOYSA-N
3-acetoxy-2-methylbenzoic acid,2-methyl-3-acetoxybenzoic acid,3-acetoxy-2-methyl benzoic acid,3-acetyloxy-2-methylbenzoic acid,benzoic acid, 3-acetyloxy-2-methyl,3-acetoxy-2-methyl-benzoic acid,acmc-209dz2,ksc495o2n,3-acetyloxy-2-methyl-benzoic acid
3798557
3-acetyloxy-2-methylbenzoic acid
CC1=C(C=CC=C1OC(=O)C)C(=O)O
168899-58-9
C10H10O4
194.19
LHVJUPHCLWIPLZ-UHFFFAOYSA-N
3-acetoxy-2-methylbenzoic acid,2-methyl-3-acetoxybenzoic acid,3-acetoxy-2-methyl benzoic acid,3-acetyloxy-2-methylbenzoic acid,benzoic acid, 3-acetyloxy-2-methyl,3-acetoxy-2-methyl-benzoic acid,acmc-209dz2,ksc495o2n,3-acetyloxy-2-methyl-benzoic acid
3798557
3-acetyloxy-2-methylbenzoic acid
CC1=C(C=CC=C1OC(=O)C)C(=O)O
Melting Point 148°C to 151°C
Color Brown to White
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 194.19
Physical Form Crystalline Powder or Powder
Percent Purity 98%
Chemical Name or Material 3-Acetoxy-2-methylbenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning