Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-acetoxybenzofuran, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
93680-80-9
C10H8O3
176.171
MFCD00151788
XCBLZAJCKKRBED-UHFFFAOYSA-N
3-acetoxybenzo b furan,3-acetoxybenzofuran,benzofuran-3-yl acetate,acmc-20anaw,3-acetoxyl-benzofuran,pubchem16309,benzo b furan-3-yl acetate,1-benzofuran-3-yl ethanoate,acetic acid 3-benzofuranyl ester
2769621
1-benzofuran-3-yl acetate
CC(=O)OC1=COC2=CC=CC=C21
93680-80-9
C10H8O3
176.171
MFCD00151788
XCBLZAJCKKRBED-UHFFFAOYSA-N
3-acetoxybenzo b furan,3-acetoxybenzofuran,benzofuran-3-yl acetate,acmc-20anaw,3-acetoxyl-benzofuran,pubchem16309,benzo b furan-3-yl acetate,1-benzofuran-3-yl ethanoate,acetic acid 3-benzofuranyl ester
2769621
1-benzofuran-3-yl acetate
CC(=O)OC1=COC2=CC=CC=C21
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Refractive Index 1.543 to 1.545 (20°C, 589nm)
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 176.17
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 3-Acetoxybenzofuran
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.