Alfa Aesar™ 3-Acetyl-1-(phenylsulfonyl)pyrrole, 98%

Manufacturer: Alfa Aesar™ H5575114

Quantity 25g

 View more versions of this product

Catalog No. AAH5575114

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 81453-98-7
    Formula Weight 249.28
    MDL Number MFCD00067752
    Chemical Name or Material 3-Acetyl-1-(phenylsulfonyl)pyrrole

    Specifications

    Formula Weight 249.28
    Melting Point 96° to 99°C
    Physical Form Solid
    MDL Number MFCD00067752
    CAS 81453-98-7
    Quantity 25g
    Chemical Name or Material 3-Acetyl-1-(phenylsulfonyl)pyrrole
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit