Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Acetylbenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
204841-19-0
C8H9BO3
163.967
MFCD01074678
SJGGDZCTGBKBCK-UHFFFAOYSA-N
3-acetylphenyl boronic acid,3-acetylbenzeneboronic acid,3-acetylphenylboronicacid,m-acetylphenylboronic acid,3'-boronoacetophenone,boronic acid, 3-acetylphenyl,1-3-dihydroxyboranyl phenyl ethan-1-one,pubchem2751,acmc-209faq,3-acetylphenyboronic acid
2734310
CHEBI:86552
(3-acetylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)C)(O)O
204841-19-0
C8H9BO3
163.967
MFCD01074678
SJGGDZCTGBKBCK-UHFFFAOYSA-N
3-acetylphenyl boronic acid,3-acetylbenzeneboronic acid,3-acetylphenylboronicacid,m-acetylphenylboronic acid,3'-boronoacetophenone,boronic acid, 3-acetylphenyl,1-3-dihydroxyboranyl phenyl ethan-1-one,pubchem2751,acmc-209faq,3-acetylphenyboronic acid
2734310
CHEBI:86552
(3-acetylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)C)(O)O
Melting Point ~200°C to 205°C
Quantity 25g
Beilstein 8255673
Formula Weight 163.97
Percent Purity 97%
Chemical Name or Material 3-Acetylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning