Alfa Aesar™ 3-Acetylbenzo[b]thiophene, 98%

Manufacturer: Alfa Aesar™ A1033014

Quantity 25g

 View more versions of this product

Catalog No. AAA1033014

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1128-05-8
    Molecular Formula C10H8OS
    Formula Weight 176.23
    MDL Number MFCD00051638
    Beilstein 122587
    Synonym 3-Acetylthianaphthene
    Chemical Name or Material 3-Acetylbenzo[b]thiophene

    Specifications

    Formula Weight 176.23
    Molecular Formula C10H8OS
    Beilstein 122587
    Synonym 3-Acetylthianaphthene
    MDL Number MFCD00051638
    CAS 1128-05-8
    Boiling Point 165° to 170°C (10mmHg)
    Melting Point 61° to 66°C
    Quantity 25g
    RTECS Number OB6125000
    Chemical Name or Material 3-Acetylbenzo[b]thiophene
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit