Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Acetylphenoxy)acetic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
1878-80-4
C10H10O4
194.186
MFCD02180947
VDZJGFDUSYCVJA-UHFFFAOYSA-N
241887
2-(3-acetylphenoxy)acetic acid
CC(=O)C1=CC(=CC=C1)OCC(=O)O
1878-80-4
C10H10O4
194.186
MFCD02180947
VDZJGFDUSYCVJA-UHFFFAOYSA-N
241887
2-(3-acetylphenoxy)acetic acid
CC(=O)C1=CC(=CC=C1)OCC(=O)O
Melting Point 119°C
Color White
Quantity 25g
Formula Weight 194.19
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (3-Acetylphenoxy)acetic Acid