Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetylphenylboronic acid, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
204841-19-0
C8H9BO3
163.97
MFCD01074678
SJGGDZCTGBKBCK-UHFFFAOYSA-N
3-acetylphenyl boronic acid, 3-acetylbenzeneboronic acid, 3-acetylphenylboronicacid, m-acetylphenylboronic acid, 3'-boronoacetophenone, boronic acid, 3-acetylphenyl, 1-3-dihydroxyboranyl phenyl ethan-1-one, pubchem2751, acmc-209faq, 3-acetylphenyboronic acid
2734310
CHEBI:86552
(3-acetylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)C)(O)O
204841-19-0
C8H9BO3
163.97
MFCD01074678
SJGGDZCTGBKBCK-UHFFFAOYSA-N
3-acetylphenyl boronic acid, 3-acetylbenzeneboronic acid, 3-acetylphenylboronicacid, m-acetylphenylboronic acid, 3'-boronoacetophenone, boronic acid, 3-acetylphenyl, 1-3-dihydroxyboranyl phenyl ethan-1-one, pubchem2751, acmc-209faq, 3-acetylphenyboronic acid
2734310
CHEBI:86552
(3-acetylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)C)(O)O
Color Yellow
Melting Point 212°C to 214°C
Packaging Glass bottle
Quantity 10g
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (HPLC)
Formula Weight 163.97
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material 3-Acetylphenylboronic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning