Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Click to view available options
Quantity:
5g
204841-19-0
C8H9BO3
163.967
MFCD01074678
SJGGDZCTGBKBCK-UHFFFAOYSA-N
3-acetylphenyl boronic acid,3-acetylbenzeneboronic acid,3-acetylphenylboronicacid,m-acetylphenylboronic acid,3'-boronoacetophenone,boronic acid, 3-acetylphenyl,1-3-dihydroxyboranyl phenyl ethan-1-one,pubchem2751,acmc-209faq,3-acetylphenyboronic acid
2734310
CHEBI:86552
(3-acetylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)C)(O)O
204841-19-0
C8H9BO3
163.967
MFCD01074678
SJGGDZCTGBKBCK-UHFFFAOYSA-N
3-acetylphenyl boronic acid,3-acetylbenzeneboronic acid,3-acetylphenylboronicacid,m-acetylphenylboronic acid,3'-boronoacetophenone,boronic acid, 3-acetylphenyl,1-3-dihydroxyboranyl phenyl ethan-1-one,pubchem2751,acmc-209faq,3-acetylphenyboronic acid
2734310
CHEBI:86552
(3-acetylphenyl)boronic acid
B(C1=CC(=CC=C1)C(=O)C)(O)O
Melting Point 208°C
Color Yellow
Quantity 5g
Formula Weight 163.97
Physical Form Crystal-Powder at 20°C
Chemical Name or Material 3-Acetylphenylboronic Acid (contains varying amounts of Anhydride)