Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetylthianaphthene, 98%, Acros Organics

Click to view available options
Quantity:
1g
1128-05-8
C10H8OS
176.233
ZTTZKDDWXHQKSY-UHFFFAOYSA-N
3-acetylthianaphthene,3-acetylbenzothiophene,3-acetylthionaphthene,3-acetylbenzo b thiophene,1-benzo b thiophen-3-yl ethanone,3-acetylbenzo b thiophene,ketone, methyl 3-thianaphthenyl,3-acetyl benzothiophene,ethanone, 1-benzo b thien-3-yl,ethanone, 1-benzo b thien-3-yl
14316
1-(1-benzothiophen-3-yl)ethanone
CC(=O)C1=CSC2=CC=CC=C21
1128-05-8
C10H8OS
176.233
ZTTZKDDWXHQKSY-UHFFFAOYSA-N
3-acetylthianaphthene,3-acetylbenzothiophene,3-acetylthionaphthene,3-acetylbenzo b thiophene,1-benzo b thiophen-3-yl ethanone,3-acetylbenzo b thiophene,ketone, methyl 3-thianaphthenyl,3-acetyl benzothiophene,ethanone, 1-benzo b thien-3-yl,ethanone, 1-benzo b thien-3-yl
14316
1-(1-benzothiophen-3-yl)ethanone
CC(=O)C1=CSC2=CC=CC=C21
Quantity 1g
Infrared Spectrum Authentic
Color Yellow
Melting Point 61°C to 65°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 17, V,10, 173
Formula Weight 176.24
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Acetylthianaphthene
Show More Show Less