Alfa Aesar™ 3-Acetylthiophene, 98%

Manufacturer: Alfa Aesar™ A1107818

Quantity 50g

 View more versions of this product

Catalog No. AAA1107818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1468-83-3
    Molecular Formula C6H6OS
    Formula Weight 126.18
    MDL Number MFCD00005468
    UN Number UN2811
    Beilstein 107241
    Synonym Methyl 3-thienyl ketone
    Chemical Name or Material 3-Acetylthiophene

    Specifications

    Formula Weight 126.18
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H6OS
    CAS 1468-83-3
    UN Number UN2811
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Quantity 50g
    Recommended Storage 2°C to 8°C
    MDL Number MFCD00005468
    Synonym Methyl 3-thienyl ketone
    Melting Point 58° to 61°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Beilstein 107241
    Boiling Point 208° to 210°C
    Chemical Name or Material 3-Acetylthiophene
    Assay 98%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    2°C to 8°C

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit