Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetylthiophene, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
1468-83-3
C6H6OS
126.18
MFCD00005468
RNIDWJDZNNVFDY-UHFFFAOYSA-N
3-acetylthiophene,methyl 3-thienyl ketone,1-thiophen-3-yl ethan-1-one,1-thiophen-3-yl ethanone,1-3-thienyl ethanone,ethanone, 1-3-thienyl,ketone, methyl 3-thienyl,3-acetyl thiophene,1-thien-3-ylethanone,methyl-3-thienyl ketone
15116
1-thiophen-3-ylethanone
CC(=O)C1=CSC=C1
1468-83-3
C6H6OS
126.18
MFCD00005468
RNIDWJDZNNVFDY-UHFFFAOYSA-N
3-acetylthiophene,methyl 3-thienyl ketone,1-thiophen-3-yl ethan-1-one,1-thiophen-3-yl ethanone,1-3-thienyl ethanone,ethanone, 1-3-thienyl,ketone, methyl 3-thienyl,3-acetyl thiophene,1-thien-3-ylethanone,methyl-3-thienyl ketone
15116
1-thiophen-3-ylethanone
CC(=O)C1=CSC=C1
Boiling Point 208°C to 210°C (748.0mmHg)
Color Beige to Brown
Infrared Spectrum Authentic
Melting Point 57°C to 61°C
Quantity 10g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C4H3S)COCH3
Beilstein 17,III,452
Formula Weight 126.18
Physical Form Adhering Crystals
Percent Purity 98%
Chemical Name or Material 3-Acetylthiophene, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-996-4

TSCA : TSCA