Alfa Aesar™ 3-Allylsalicylaldehyde, 97%

Manufacturer: Alfa Aesar™ H5347514

Quantity 25g

 View more versions of this product

Catalog No. AAH5347514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24019-66-7
    Molecular Formula C10H10O2
    Formula Weight 162.19
    MDL Number MFCD00037364
    Synonym 3-Allyl-2-hydroxybenzaldehyde
    Chemical Name or Material 3-Allylsalicylaldehyde

    Specifications

    Formula Weight 162.19
    Quantity 25g
    Flash Point 107°C (225°F)
    Molecular Formula C10H10O2
    MDL Number MFCD00037364
    CAS 24019-66-7
    Synonym 3-Allyl-2-hydroxybenzaldehyde
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 3-Allylsalicylaldehyde
    Assay 97%

    Description

    Applications
    3-Allylsalicylaldehyde is a synthetic intermediate for the synthesis of 3-aminoflavones. Flavonoids have been studied for their antiproliferative activity and in vitro cytotoxicity. It is used in the preparation of oxazole derivatives and also imidazole scaffold-based 2-substituted benzofurans for application against human tumor cell lines. It is a reagent in the synthesis of antioxidant and antimicrobial novel chalcones.

    Solubility
    Soluble in chloroform.

    Notes
    Air sensitive. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit