Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-1-(2,4,6-trichlorophenyl)-5-pyrazolone , 97%

Click to view available options
Quantity:
250mg
1g
27241-31-2
C9H6Cl3N3O
278.517
MFCD00020743
FOGGKKWSFIESMZ-UHFFFAOYSA-N
3-amino-1-2,4,6-trichlorophenyl-1h-pyrazol-5 4h-one,3-amino-1-2,4,6-trichlorophenyl-2-pyrazolin-5-one,3-amino-1-2,4,6-trichlorophenyl-5-pyrazolone,5-amino-2-2,4,6-trichlorophenyl-4h-pyrazol-3-one,1-2,4,6-trichlorophenyl-3-anilinopyrazolone,3h-pyrazol-3-one, 5-amino-2,4-dihydro-2-2,4,6-trichlorophenyl,5-amino-2-2,4,6-trichlorophenyl-2,4-dihydro-3h-pyrazol-3-one,1-2,4,6-trichlorophenyl 3-amino-5-pyrazolone,3-amino-1-2,4,6-trichlorophenyl-4,5-dihydro-1h-pyrazol-5-one,1-2,4,6-trichlorophenyl-3-amino-5-pyrazolone
270746
5-amino-2-(2,4,6-trichlorophenyl)-4H-pyrazol-3-one
C1C(=NN(C1=O)C2=C(C=C(C=C2Cl)Cl)Cl)N
27241-31-2
C9H6Cl3N3O
278.517
MFCD00020743
FOGGKKWSFIESMZ-UHFFFAOYSA-N
3-amino-1-2,4,6-trichlorophenyl-1h-pyrazol-5 4h-one,3-amino-1-2,4,6-trichlorophenyl-2-pyrazolin-5-one,3-amino-1-2,4,6-trichlorophenyl-5-pyrazolone,5-amino-2-2,4,6-trichlorophenyl-4h-pyrazol-3-one,1-2,4,6-trichlorophenyl-3-anilinopyrazolone,3h-pyrazol-3-one, 5-amino-2,4-dihydro-2-2,4,6-trichlorophenyl,5-amino-2-2,4,6-trichlorophenyl-2,4-dihydro-3h-pyrazol-3-one,1-2,4,6-trichlorophenyl 3-amino-5-pyrazolone,3-amino-1-2,4,6-trichlorophenyl-4,5-dihydro-1h-pyrazol-5-one,1-2,4,6-trichlorophenyl-3-amino-5-pyrazolone
270746
5-amino-2-(2,4,6-trichlorophenyl)-4H-pyrazol-3-one
C1C(=NN(C1=O)C2=C(C=C(C=C2Cl)Cl)Cl)N
Quantity 250mg
Beilstein 535761
Formula Weight 278.52
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Amino-1-(2,4,6-trichlorophenyl)-5-pyrazolone