Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1,2,4-triazine, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1120-99-6
C3H4N4
96.09
MFCD00006459
MJIWQHRXSLOUJN-UHFFFAOYSA-N
3-amino-1,2,4-triazine,as-triazine, 3-amino,3-amino-as-triazine,1,2,4-triazin-3-ylamine,1,2,4-triazine-3-ylamine,1,2,4 triazin-3-ylamine,zlchem 1277,1,2,4-triazine-3-amine,3-amino-1,2,4,-triazine
70715
1,2,4-triazin-3-amine
C1=CN=NC(=N1)N
1120-99-6
C3H4N4
96.09
MFCD00006459
MJIWQHRXSLOUJN-UHFFFAOYSA-N
3-amino-1,2,4-triazine,as-triazine, 3-amino,3-amino-as-triazine,1,2,4-triazin-3-ylamine,1,2,4-triazine-3-ylamine,1,2,4 triazin-3-ylamine,zlchem 1277,1,2,4-triazine-3-amine,3-amino-1,2,4,-triazine
70715
1,2,4-triazin-3-amine
C1=CN=NC(=N1)N
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 175°C to 177°C
Color Brown
Quantity 10g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 96.09
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Amino-1, 2, 4-triazine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-324-7