Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-3-Amino-1,2-propanediol, 97%

Click to view available options
Quantity:
25g
100g
500g
616-30-8
C3H9NO2
91.11
MFCD00008140
KQIGMPWTAHJUMN-UHFFFAOYSA-N
3-amino-1,2-propanediol,2,3-dihydroxypropylamine,1,2-propanediol, 3-amino,1-amino-2,3-propanediol,1-aminoglycerol,2,3-propandiol-1-amine,1-aminopropanediol,3-aminopropan-1,2-diol,3-amino-1,2-proopanediol,+/--3-amino-1,2-propanediol
73561
3-aminopropane-1,2-diol
C(C(CO)O)N
616-30-8
C3H9NO2
91.11
MFCD00008140
KQIGMPWTAHJUMN-UHFFFAOYSA-N
3-amino-1,2-propanediol,2,3-dihydroxypropylamine,1,2-propanediol, 3-amino,1-amino-2,3-propanediol,1-aminoglycerol,2,3-propandiol-1-amine,1-aminopropanediol,3-aminopropan-1,2-diol,3-amino-1,2-proopanediol,+/--3-amino-1,2-propanediol
73561
3-aminopropane-1,2-diol
C(C(CO)O)N
Applications
3-Amino-1,2-propanediol is used in the preparation of iohexol, which is a X-CT non-ionic contrast medium. It acts as a starting material in the production of specialty materials.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
Quantity 100g
Flash Point 186°C (366°F)
Boiling Point 264°C to 266°C (739mmHg)
UN Number UN2735
Beilstein 1719121
Formula Weight 91.11
Percent Purity 97%
Chemical Name or Material (+/-)-3-Amino-1,2-propanediol

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : TY2800000

TSCA : TSCA