Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1,2-propanediol 98.0+%, TCI America™

Click to view available options
Quantity:
10g
25g
616-30-8
C3H9NO2
91.11
MFCD00008140
KQIGMPWTAHJUMN-UHFFFAOYSA-N
3-amino-1,2-propanediol,2,3-dihydroxypropylamine,1,2-propanediol, 3-amino,1-amino-2,3-propanediol,1-aminoglycerol,2,3-propandiol-1-amine,1-aminopropanediol,3-aminopropan-1,2-diol,3-amino-1,2-proopanediol,+/--3-amino-1,2-propanediol
73561
3-aminopropane-1,2-diol
C(C(CO)O)N
616-30-8
C3H9NO2
91.11
MFCD00008140
KQIGMPWTAHJUMN-UHFFFAOYSA-N
3-amino-1,2-propanediol,2,3-dihydroxypropylamine,1,2-propanediol, 3-amino,1-amino-2,3-propanediol,1-aminoglycerol,2,3-propandiol-1-amine,1-aminopropanediol,3-aminopropan-1,2-diol,3-amino-1,2-proopanediol,+/--3-amino-1,2-propanediol
73561
3-aminopropane-1,2-diol
C(C(CO)O)N
Color Yellow
Quantity 10g
UN Number 2735
Formula Weight 91.11
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3-Amino-1,2-propanediol

EINECSNumber : (2)-3504

RTECSNumber : TY2800000

TSCA : Yes