Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1,5-naphthalenedisulfonic Acid Disodium Salt 98.0+%, TCI America™

Click to view available options
Quantity:
25g
14170-43-5
C10H7NNa2O6S2
347.267
MFCD00070487
UZUODNWWWUQRIR-UHFFFAOYSA-L
C Acid, 2-Naphthylamine-4,8-disulfonic Acid Disodium Salt, Disodium 3-Amino-1,5-naphthalenedisulfonate
84232
disodium;3-aminonaphthalene-1,5-disulfonate
C1=CC2=C(C=C(C=C2C(=C1)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
14170-43-5
C10H7NNa2O6S2
347.267
MFCD00070487
UZUODNWWWUQRIR-UHFFFAOYSA-L
C Acid, 2-Naphthylamine-4,8-disulfonic Acid Disodium Salt, Disodium 3-Amino-1,5-naphthalenedisulfonate
84232
disodium;3-aminonaphthalene-1,5-disulfonate
C1=CC2=C(C=C(C=C2C(=C1)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
Color Undesignated
Quantity 25g
Formula Weight 347.27
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material 3-Amino-1,5-naphthalenedisulfonic Acid Disodium Salt

EINECSNumber : (4)-0502

TSCA : No