Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-methyl-1H-indazole, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
60301-20-4
C8H9N3
147.18
MFCD09054755
NYLGITXFVVEBLZ-UHFFFAOYSA-N
1-methyl-1h-indazol-3-amine,3-amino-1-methylindazole,3-amino-1-methyl-1h-indazole,1h-indazol-3-amine, 1-methyl,n-methyl-3-aminoindazole,1-methyl-1h-3-indazoleamine,1-methyl-1h-indazole-3-amine,1-methyl-1h-indazol-3-ylamine,1-methyl-1h-indazole-3-ylamine
12291317
1-methylindazol-3-amine
CN1C2=CC=CC=C2C(=N1)N
60301-20-4
C8H9N3
147.18
MFCD09054755
NYLGITXFVVEBLZ-UHFFFAOYSA-N
1-methyl-1h-indazol-3-amine,3-amino-1-methylindazole,3-amino-1-methyl-1h-indazole,1h-indazol-3-amine, 1-methyl,n-methyl-3-aminoindazole,1-methyl-1h-3-indazoleamine,1-methyl-1h-indazole-3-amine,1-methyl-1h-indazol-3-ylamine,1-methyl-1h-indazole-3-ylamine
12291317
1-methylindazol-3-amine
CN1C2=CC=CC=C2C(=N1)N
Melting Point 95°C to 99°C
Quantity 5g
Color Brown to White
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 147.18
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 3-Amino-1-methyl-1H-indazole
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning