Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-methyl-1H-indazole, 95%, Thermo Scientific Chemicals

Catalog Number p-4228321
Click to view available options
Quantity:
1 g
5 g
60301-20-4
C8H9N3
147.18
MFCD09054755
NYLGITXFVVEBLZ-UHFFFAOYSA-N
1-methyl-1h-indazol-3-amine, 3-amino-1-methylindazole, 3-amino-1-methyl-1h-indazole, 1h-indazol-3-amine, 1-methyl, n-methyl-3-aminoindazole, 1-methyl-1h-3-indazoleamine, 1-methyl-1h-indazole-3-amine, 1-methyl-1h-indazol-3-ylamine, 1-methyl-1h-indazole-3-ylamine
12291317
CN1N=C(N)C2=CC=CC=C12
60301-20-4
C8H9N3
147.18
MFCD09054755
NYLGITXFVVEBLZ-UHFFFAOYSA-N
1-methyl-1h-indazol-3-amine, 3-amino-1-methylindazole, 3-amino-1-methyl-1h-indazole, 1h-indazol-3-amine, 1-methyl, n-methyl-3-aminoindazole, 1-methyl-1h-3-indazoleamine, 1-methyl-1h-indazole-3-amine, 1-methyl-1h-indazol-3-ylamine, 1-methyl-1h-indazole-3-ylamine
12291317
CN1N=C(N)C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 95°C to 99°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
IUPAC Name 1-methyl-1H-indazol-3-amine
Formula Weight 147.18
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1-methyl-1H-indazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit