Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-methyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
886851-66-7
C5H7N3O
125.13
MFCD00460466
UGWUYEXHMDPAMR-UHFFFAOYSA-N
3-amino-1-methyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3-amino-1-methyl, 1h-pyrazole-4-carboxaldehyde, 3-amino-1-methyl, 3-amino-1-methyl-1h-pyrazole-4-carboxaldehyde
18525942
3-amino-1-methylpyrazole-4-carbaldehyde
CN1C=C(C=O)C(N)=N1
886851-66-7
C5H7N3O
125.13
MFCD00460466
UGWUYEXHMDPAMR-UHFFFAOYSA-N
3-amino-1-methyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3-amino-1-methyl, 1h-pyrazole-4-carboxaldehyde, 3-amino-1-methyl, 3-amino-1-methyl-1h-pyrazole-4-carboxaldehyde
18525942
3-amino-1-methylpyrazole-4-carbaldehyde
CN1C=C(C=O)C(N)=N1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material 3-Amino-1-methyl-1H-pyrazole-4-carbaldehyde

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning