Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-methyl-5-(trifluoromethyl)-1H-indazole, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
5685-69-8
C9H8F3N3
215.18
MFCD07777192
IRSFDHYOPSXICW-UHFFFAOYSA-N
1-methyl-5-trifluoromethyl-1h-indazol-3-amine,1-methyl-5-trifluoromethyl indazol-3-amine,1-methyl-5-trifluoromethyl-1h-indazol-3-ylamine,3-amino-5-trifluoromethyl-1-methyl-indazole,3-amino-1-methyl-5-trifluoromethyl-1h-indazole,1-methyl-5-trifluoromethyl-1h-indazole-3-ylamine
28875377
1-methyl-5-(trifluoromethyl)indazol-3-amine
CN1C2=C(C=C(C=C2)C(F)(F)F)C(=N1)N
5685-69-8
C9H8F3N3
215.18
MFCD07777192
IRSFDHYOPSXICW-UHFFFAOYSA-N
1-methyl-5-trifluoromethyl-1h-indazol-3-amine,1-methyl-5-trifluoromethyl indazol-3-amine,1-methyl-5-trifluoromethyl-1h-indazol-3-ylamine,3-amino-5-trifluoromethyl-1-methyl-indazole,3-amino-1-methyl-5-trifluoromethyl-1h-indazole,1-methyl-5-trifluoromethyl-1h-indazole-3-ylamine
28875377
1-methyl-5-(trifluoromethyl)indazol-3-amine
CN1C2=C(C=C(C=C2)C(F)(F)F)C(=N1)N
Melting Point 145°C to 147°C
Quantity 5g
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 215.18
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 3-Amino-1-methyl-5-(trifluoromethyl)-1H-indazole
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning