Learn More
3-Amino-1H-1,2,4-triazole, 96%, Thermo Scientific Chemicals
Description
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.
Solubility
It is soluble in water, methanol, ethanol and chloroform, and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.
Notes
Store desiccated at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from Strong oxidizing agents, Strong acids, Copper, Iron and iron salts.
Specifications
Melting Point | 147°C to 153°C |
Density | 1.138 |
Assay Percent Range | 96% |
Quantity | 50 g |
UN Number | UN3077 |
Beilstein | 107687 |
Merck Index | 14,489 |
Solubility Information | It is soluble in water,methanol,ethanol and chloroform,and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone. |
Formula Weight | 84.08 |
Percent Purity | 96% |
Show More |
Safety and Handling
Hazard Category | H302-H319-H350-H360-H373 |
Hazard Statement | GHS H Statement H361-H373 |
Precautionary Statement | P201-P202-P260-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 200-521-5 |
RTECSNumber | XZ3850000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.