Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2,2-dimethyl-1-propanol 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
26734-09-8
C5H13NO
103.165
MFCD00059847
FNVOFDGAASRDQY-UHFFFAOYSA-N
3-amino-2,2-dimethyl-1-propanol,neopentanolamine,1-propanol, 3-amino-2,2-dimethyl,3-amino-2,2-dimethylpropanol,2,2-dimethyl-3-aminopropanol,1-amino-2,2-dimethyl-3-propanol,3-amino-2,2-dimethyl-propan-1-ol,neo-pentanolamine,acmc-1cdms,ksc496k6d
117326
3-amino-2,2-dimethylpropan-1-ol
CC(C)(CN)CO
26734-09-8
C5H13NO
103.165
MFCD00059847
FNVOFDGAASRDQY-UHFFFAOYSA-N
3-amino-2,2-dimethyl-1-propanol,neopentanolamine,1-propanol, 3-amino-2,2-dimethyl,3-amino-2,2-dimethylpropanol,2,2-dimethyl-3-aminopropanol,1-amino-2,2-dimethyl-3-propanol,3-amino-2,2-dimethyl-propan-1-ol,neo-pentanolamine,acmc-1cdms,ksc496k6d
117326
3-amino-2,2-dimethylpropan-1-ol
CC(C)(CN)CO
Color White
Quantity 25g
UN Number 3263
Formula Weight 103.17
Physical Form Crystal-Lump at 20°C
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 3-Amino-2,2-dimethyl-1-propanol