Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2,6-dibromopyridine, 97%, Thermo Scientific™

Click to view available options
Quantity:
5g
39856-57-0
C5H4Br2N2
251.91
CRTOIQFRVBJJRI-UHFFFAOYSA-N
3-amino-2,6-dibromopyridine, 2,6-dibromo-3-pyridinamine, 2,6-dibromo-pyridin-3-ylamine, 3-pyridinamine, 2,6-dibromo, 2,6-dibromo-3-pyridylamine, 2,6-dibromo-3-aminopyridine, zlchem 949, pubchem9809, acmc-209j8e, ksc495i3f
817678
2,6-dibromopyridin-3-amine
C1=CC(=NC(=C1N)Br)Br
39856-57-0
C5H4Br2N2
251.91
CRTOIQFRVBJJRI-UHFFFAOYSA-N
3-amino-2,6-dibromopyridine, 2,6-dibromo-3-pyridinamine, 2,6-dibromo-pyridin-3-ylamine, 3-pyridinamine, 2,6-dibromo, 2,6-dibromo-3-pyridylamine, 2,6-dibromo-3-aminopyridine, zlchem 949, pubchem9809, acmc-209j8e, ksc495i3f
817678
2,6-dibromopyridin-3-amine
C1=CC(=NC(=C1N)Br)Br
Quantity 5g
Melting Point 147°C to 148°C
Assay Percent Range 96% min. (GC)
Formula Weight 251.91
Percent Purity 97%
Chemical Name or Material 3-Amino-2, 6-dibromopyridine

GHS Signal Word: Warning

RUO – Research Use Only