Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-2,6-difluorobenzoic acid, 95%

Click to view available options
Quantity:
1g
5g
83141-11-1
C7H5F2NO2
173.119
MFCD07368833
ORSUTLAFOCNIDY-UHFFFAOYSA-N
3-amino-2,6-difluorobenzoicacid,pubchem8200,3-amino-2,6-difluoro-benzoic acid,buttpark 120\01-13,benzoic acid,3-amino-2,6-difluoro,benzoic acid, 3-amino-2,6-difluoro
18405352
3-amino-2,6-difluorobenzoic acid
C1=CC(=C(C(=C1N)F)C(=O)O)F
83141-11-1
C7H5F2NO2
173.119
MFCD07368833
ORSUTLAFOCNIDY-UHFFFAOYSA-N
3-amino-2,6-difluorobenzoicacid,pubchem8200,3-amino-2,6-difluoro-benzoic acid,buttpark 120\01-13,benzoic acid,3-amino-2,6-difluoro,benzoic acid, 3-amino-2,6-difluoro
18405352
3-amino-2,6-difluorobenzoic acid
C1=CC(=C(C(=C1N)F)C(=O)O)F
Quantity 5g
Beilstein 13330504
Formula Weight 173.12
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-Amino-2,6-difluorobenzoic acid