Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2,7-naphthalenedisulfonic Acid Monosodium Salt 90.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
5332-41-2
C10H9NNaO6S2
326.293
MFCD00004114
YMVVPNVDLFEING-UHFFFAOYSA-N
Amino R Acid Monosodium Salt, 2-Naphthylamine-3,6-disulfonic Acid Monosodium Salt, Monosodium 3-Amino-2,7-naphthalenedisulfonate, Sodium Hydrogen 3-Amino-2,7-naphthalenedisulfonate
88804670
3-aminonaphthalene-2,7-disulfonic acid;sodium
C1=CC(=CC2=CC(=C(C=C21)N)S(=O)(=O)O)S(=O)(=O)O.[Na]
5332-41-2
C10H9NNaO6S2
326.293
MFCD00004114
YMVVPNVDLFEING-UHFFFAOYSA-N
Amino R Acid Monosodium Salt, 2-Naphthylamine-3,6-disulfonic Acid Monosodium Salt, Monosodium 3-Amino-2,7-naphthalenedisulfonate, Sodium Hydrogen 3-Amino-2,7-naphthalenedisulfonate
88804670
3-aminonaphthalene-2,7-disulfonic acid;sodium
C1=CC(=CC2=CC(=C(C=C21)N)S(=O)(=O)O)S(=O)(=O)O.[Na]
Color Undesignated
Quantity 500g
Formula Weight 325.28
Physical Form Crystal-Powder at 20°C
Percent Purity ≥90.0% (T)
Chemical Name or Material 3-Amino-2,7-naphthalenedisulfonic Acid Monosodium Salt

EINECSNumber : (4)-0502

TSCA : No