Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-2-bromo-6-methylpyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
126325-48-2
C6H7BrN2
187.04
MFCD03095078
ORELOTUWTIKRIW-UHFFFAOYSA-N
3-amino-4-bromo-2-methylpyridine,ksc915a1f,3-pyridinamine, 4-bromo-2-methyl,4-bromo-2-methyl-3-pyridinamine
14418044
4-bromo-2-methylpyridin-3-amine
CC1=NC=CC(=C1N)Br
126325-48-2
C6H7BrN2
187.04
MFCD03095078
ORELOTUWTIKRIW-UHFFFAOYSA-N
3-amino-4-bromo-2-methylpyridine,ksc915a1f,3-pyridinamine, 4-bromo-2-methyl,4-bromo-2-methyl-3-pyridinamine
14418044
4-bromo-2-methylpyridin-3-amine
CC1=NC=CC(=C1N)Br
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Quantity 1g
Color White
UN Number UN2811
Beilstein 4243986
Formula Weight 187.04
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-Amino-2-bromo-6-methylpyridine

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C