Alfa Aesar™ 3-Amino-2-chloro-4-methylpyridine, 97%

Manufacturer: Alfa Aesar™ H2605806

Quantity 5g

 View more versions of this product

Catalog No. AAH2605806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 133627-45-9
    Molecular Formula C6H7ClN2
    Formula Weight 142.59
    MDL Number MFCD00673152
    Synonym 3-Amino-2-chloro-4-picoline; 2-Chloro-4-methyl-3-pyridinamine
    Chemical Name or Material 3-Amino-2-chloro-4-methylpyridine

    Specifications

    Formula Weight 142.59
    Melting Point 64° to 68°C
    Quantity 5g
    Molecular Formula C6H7ClN2
    MDL Number MFCD00673152
    Synonym 3-Amino-2-chloro-4-picoline; 2-Chloro-4-methyl-3-pyridinamine
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 133627-45-9
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 3-Amino-2-chloro-4-methylpyridine
    Assay 97%

    GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit