Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-2-chloro-5-(trifluoromethyl)pyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
72587-18-9
C6H4ClF3N2
196.557
MFCD07375383
HABKYPVMMHBOIW-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl pyridin-3-amine,2-chloro-5-trifluoromethyl-3-pyridinamine,3-amino-2-chloro-5-trifluoromethyl pyridine,3-amino-2-chloro-5-trifluoromethylpyridine,2-chloro-3-amino-5-trifluoromethyl pyridine
12919757
2-chloro-5-(trifluoromethyl)pyridin-3-amine
C1=C(C=NC(=C1N)Cl)C(F)(F)F
72587-18-9
C6H4ClF3N2
196.557
MFCD07375383
HABKYPVMMHBOIW-UHFFFAOYSA-N
2-chloro-5-trifluoromethyl pyridin-3-amine,2-chloro-5-trifluoromethyl-3-pyridinamine,3-amino-2-chloro-5-trifluoromethyl pyridine,3-amino-2-chloro-5-trifluoromethylpyridine,2-chloro-3-amino-5-trifluoromethyl pyridine
12919757
2-chloro-5-(trifluoromethyl)pyridin-3-amine
C1=C(C=NC(=C1N)Cl)C(F)(F)F
Applications
It’s employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Quantity 1g
Color White to Brown
Beilstein 19757170
Formula Weight 196.56
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-Amino-2-chloro-5-(trifluoromethyl)pyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C