Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-2-chloropyridine, 98+%

Click to view available options
Quantity:
10g
50g
250g
6298-19-7
C5H5ClN2
128.559
MFCD00006238
MEQBJJUWDCYIAB-UHFFFAOYSA-N
3-amino-2-chloropyridine,2-chloro-3-aminopyridine,2-chloro-3-pyridinamine,3-pyridinamine, 2-chloro,2-chloro-3-amino pyridine,2-chloro-3-pyridylamine,2-chloro-3-amino-pyridine,2-chloro-pyridin-3-ylamine,2-chloro-3-pyridinylamine,3-amino-2-chloro-pyridine
80528
2-chloropyridin-3-amine
C1=CC(=C(N=C1)Cl)N
6298-19-7
C5H5ClN2
128.559
MFCD00006238
MEQBJJUWDCYIAB-UHFFFAOYSA-N
3-amino-2-chloropyridine,2-chloro-3-aminopyridine,2-chloro-3-pyridinamine,3-pyridinamine, 2-chloro,2-chloro-3-amino pyridine,2-chloro-3-pyridylamine,2-chloro-3-amino-pyridine,2-chloro-pyridin-3-ylamine,2-chloro-3-pyridinylamine,3-amino-2-chloro-pyridine
80528
2-chloropyridin-3-amine
C1=CC(=C(N=C1)Cl)N
Applications
It is an intermediate of medicine and agricultural chemicals. It is involved in the Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.

Solubility
Soluble in methanol and water. (30 g/L).

Notes
Store at room temperature. Keep away from oxidizing agents.
Quantity 10g
Boiling Point 130°C to 134°C (22mmHg)
Odor Unpleasant
Flash Point 185°C (365°F)
Melting Point 76°C to 80°C
Beilstein 109814
Solubility Information Soluble in methanol and water. (30g/L).
Formula Weight 128.56
Percent Purity ≥98%
Chemical Name or Material 3-Amino-2-chloropyridine

GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 228-572-9

TSCA : No

Recommended Storage : Ambient temperatures