Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-2-fluoro-4-methylpyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
173435-32-0
C6H7FN2
126.134
MFCD04112556
ZKQQKPXTKXTEQR-UHFFFAOYSA-N
3-amino-2-fluoro-4-picoline,3-amino-2-fluoro-4-methylpyridine,2-fluoro-4-methyl-pyridin-3-ylamine,2-fluoro-4-methyl-3-pyridinamine,2-fluoro-4-methyl-3-pyridylamine,3-pyridinamine, 2-fluoro-4-methyl,pubchem2984,3-amino-2-fluoro-4-methylpyridin,3-pyridinamine,2-fluoro-4-methyl,2-fluoranyl-4-methyl-pyridin-3-amine
2763161
2-fluoro-4-methylpyridin-3-amine
CC1=C(C(=NC=C1)F)N
173435-32-0
C6H7FN2
126.134
MFCD04112556
ZKQQKPXTKXTEQR-UHFFFAOYSA-N
3-amino-2-fluoro-4-picoline,3-amino-2-fluoro-4-methylpyridine,2-fluoro-4-methyl-pyridin-3-ylamine,2-fluoro-4-methyl-3-pyridinamine,2-fluoro-4-methyl-3-pyridylamine,3-pyridinamine, 2-fluoro-4-methyl,pubchem2984,3-amino-2-fluoro-4-methylpyridin,3-pyridinamine,2-fluoro-4-methyl,2-fluoranyl-4-methyl-pyridin-3-amine
2763161
2-fluoro-4-methylpyridin-3-amine
CC1=C(C(=NC=C1)F)N
Melting Point 63°C to 64°C
Quantity 250mg
Color White
Beilstein 13755262
Formula Weight 126.13
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-Amino-2-fluoro-4-methylpyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning