Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-2-methoxypyridine-4-carboxylic acid, 95%

Click to view available options
Quantity:
250mg
1g
870997-81-2
C7H8N2O3
168.152
MFCD18262266
ABVDQGNEDSKBLA-UHFFFAOYSA-N
3-amino-2-methoxyisonicotinic acid,3-amino-2-methoxy-4-pyridinecarboxylic acid,3-amino-2-methoxyisonicotinicacid,3-amino-2-methoxy-isonicotinic acid
53634618
3-amino-2-methoxypyridine-4-carboxylic acid
COC1=NC=CC(=C1N)C(=O)O
870997-81-2
C7H8N2O3
168.152
MFCD18262266
ABVDQGNEDSKBLA-UHFFFAOYSA-N
3-amino-2-methoxyisonicotinic acid,3-amino-2-methoxy-4-pyridinecarboxylic acid,3-amino-2-methoxyisonicotinicacid,3-amino-2-methoxy-isonicotinic acid
53634618
3-amino-2-methoxypyridine-4-carboxylic acid
COC1=NC=CC(=C1N)C(=O)O
Quantity 1g
Beilstein 19162520
Formula Weight 168.15
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-Amino-2-methoxypyridine-4-carboxylic acid