Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-methylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
882678-96-8
C13H20BNO2
233.12
JMKMGPGFYMANCA-UHFFFAOYSA-N
2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,3-amino-2-methylphenylboronic acid, pinacol ester,3-amino-2-methylphenylboronic acid pinacol ester,2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine,3-amino-2-methylphenyl boronic acid pinacol ester,2-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine,2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylamine,2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine,2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine
46738005
2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)N)C
882678-96-8
C13H20BNO2
233.12
JMKMGPGFYMANCA-UHFFFAOYSA-N
2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,3-amino-2-methylphenylboronic acid, pinacol ester,3-amino-2-methylphenylboronic acid pinacol ester,2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine,3-amino-2-methylphenyl boronic acid pinacol ester,2-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine,2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylamine,2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine,2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine
46738005
2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)N)C
Packaging Glass Bottle
Boiling Point 120°C to 121°C
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 233.12
Percent Purity 97%
Chemical Name or Material 3-Amino-2-methylphenylboronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning