Alfa Aesar™ 3-Amino-2-naphthol, 97%

Manufacturer: Alfa Aesar™ B2457014

Quantity 25g

 View more versions of this product

Catalog No. AAB2457014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5417-63-0
    Molecular Formula C10H9NO
    Formula Weight 159.19
    MDL Number MFCD00004113
    Synonym 2-Amino-3-hydroxynaphthalene; 3-Hydroxy-2-naphthylamine
    Chemical Name or Material 3-Amino-2-naphthol

    Specifications

    Formula Weight 159.19
    Melting Point 232° to 238°C
    Quantity 25g
    Molecular Formula C10H9NO
    Odor Odorless
    MDL Number MFCD00004113
    Synonym 2-Amino-3-hydroxynaphthalene; 3-Hydroxy-2-naphthylamine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 5417-63-0
    Chemical Name or Material 3-Amino-2-naphthol
    Assay 97%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit