Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-2-naphthol, 97%

Click to view available options
Quantity:
1g
5g
25g
5417-63-0
C10H9NO
159.188
MFCD00004113
ZHVPTERSBUMMHK-UHFFFAOYSA-N
3-amino-2-naphthol,2-naphthalenol, 3-amino,2-amino-3-naphthol,3-amino-2-hydroxynaphthalene,2-naphthol, 3-amino,2-amino-3-hydroxynaphthalene,unii-29bke27d17,3-amino-naphthalen-2-ol,acmc-209ldx,3-amino-2-naphthalenol
79449
3-aminonaphthalen-2-ol
C1=CC=C2C=C(C(=CC2=C1)N)O
5417-63-0
C10H9NO
159.188
MFCD00004113
ZHVPTERSBUMMHK-UHFFFAOYSA-N
3-amino-2-naphthol,2-naphthalenol, 3-amino,2-amino-3-naphthol,3-amino-2-hydroxynaphthalene,2-naphthol, 3-amino,2-amino-3-hydroxynaphthalene,unii-29bke27d17,3-amino-naphthalen-2-ol,acmc-209ldx,3-amino-2-naphthalenol
79449
3-aminonaphthalen-2-ol
C1=CC=C2C=C(C(=CC2=C1)N)O
Odor Odorless
Melting Point 232°C to 238°C
Quantity 5g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 3-Amino-2-naphthol

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger