Alfa Aesar™ 3-Amino-2-phenylindenone, 98%

Manufacturer: Alfa Aesar™ A1139714

Quantity 25g

 View more versions of this product

Catalog No. AAA1139714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1947-47-3
    Molecular Formula C15H11NO
    Formula Weight 221.26
    MDL Number MFCD00051614
    Beilstein 1875348
    Chemical Name or Material 3-Amino-2-phenylindenone

    Specifications

    Formula Weight 221.26
    Melting Point 268° to 274°C
    Molecular Formula C15H11NO
    MDL Number MFCD00051614
    CAS 1947-47-3
    Quantity 25g
    Beilstein 1875348
    Chemical Name or Material 3-Amino-2-phenylindenone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit