Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-2-pyrazoleethanol, 96%

Click to view available options
Quantity:
5g
25g
100g
73616-27-0
C5H9N3O
127.147
MFCD08063809
IHQRJCVJAUKIEP-UHFFFAOYSA-N
5-amino-1-2-hydroxyethyl pyrazole,2-5-aminopyrazol-1-yl ethanol,5-amino-1-,a-hydroxyethyl pyrazole,2-5-amino-1h-pyrazol-1-yl ethanol,1h-pyrazole-1-ethanol, 5-amino,2-5-amino-1h-pyrazol-1-yl ethan-1-ol,5-amino-1-2-hydroxyethyl pyrazole,pubchem15051,ksc496i9l,2-5-amino-1-pyrazolyl ethanol
320281
2-(5-aminopyrazol-1-yl)ethanol
C1=C(N(N=C1)CCO)N
73616-27-0
C5H9N3O
127.147
MFCD08063809
IHQRJCVJAUKIEP-UHFFFAOYSA-N
5-amino-1-2-hydroxyethyl pyrazole,2-5-aminopyrazol-1-yl ethanol,5-amino-1-,a-hydroxyethyl pyrazole,2-5-amino-1h-pyrazol-1-yl ethanol,1h-pyrazole-1-ethanol, 5-amino,2-5-amino-1h-pyrazol-1-yl ethan-1-ol,5-amino-1-2-hydroxyethyl pyrazole,pubchem15051,ksc496i9l,2-5-amino-1-pyrazolyl ethanol
320281
2-(5-aminopyrazol-1-yl)ethanol
C1=C(N(N=C1)CCO)N
Quantity 5g
Beilstein 607610
Formula Weight 127.15
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 3-Amino-2-pyrazoleethanol

storageNote1 : 2° to 8°C