Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-3-(2-nitrophenyl)propionic acid, 98%

Click to view available options
Quantity:
5g
25g
100g
5678-48-8
C9H10N2O4
210.189
MFCD00090356
XXBOYULKNZTOMN-UHFFFAOYSA-N
3-amino-3-2-nitrophenyl propanoic acid,3-amino-3-2-nitrophenyl propionic acid,3-amino-3-2-nitro-phenyl-propionic acid,3-2-nitrophenyl-dl-beta-alanine,3-2-nitrophenyl-beta-alanine,rs-3-amino-3-2-nitrophenyl-propionic acid,maybridge1_004875,dl-beta-2-nitrophenyl alanine,beta-amino-o-nitrohydrocinnamic acid,hydrocinnamic acid, beta-amino-o-nitro
2779217
3-amino-3-(2-nitrophenyl)propanoic acid
C1=CC=C(C(=C1)C(CC(=O)O)N)[N+](=O)[O-]
5678-48-8
C9H10N2O4
210.189
MFCD00090356
XXBOYULKNZTOMN-UHFFFAOYSA-N
3-amino-3-2-nitrophenyl propanoic acid,3-amino-3-2-nitrophenyl propionic acid,3-amino-3-2-nitro-phenyl-propionic acid,3-2-nitrophenyl-dl-beta-alanine,3-2-nitrophenyl-beta-alanine,rs-3-amino-3-2-nitrophenyl-propionic acid,maybridge1_004875,dl-beta-2-nitrophenyl alanine,beta-amino-o-nitrohydrocinnamic acid,hydrocinnamic acid, beta-amino-o-nitro
2779217
3-amino-3-(2-nitrophenyl)propanoic acid
C1=CC=C(C(=C1)C(CC(=O)O)N)[N+](=O)[O-]
Applications
3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 0-5°C. Incompatible with strong oxidizing agents, acids, bases.
Quantity 100g
Formula Weight 210.19
Percent Purity 98%
Chemical Name or Material 3-Amino-3-(2-nitrophenyl)propionic acid