Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-3-(p-methoxyphenyl)-1-propanol, 98%, Acros Organics

Click to view available options
Quantity:
1g
68208-24-2
C10H15NO2
181.235
MFCD01863268
WHWMCHUIUINGOD-UHFFFAOYSA-N
3-amino-3-4-methoxyphenyl propan-1-ol,3-amino-3-4-methoxyphenyl-1-propanol,3-amino-3-4-methoxy-phenyl-propan-1-ol,3-4-methoxyphenyl-dl-beta-alaninol,dl-beta-4-methoxyphenyl alaninol,3-amino-3-p-methoxyphenyl-1-propanol,benzenepropanol, gamma-amino-4-methoxy,1r-3-hydroxy-1-4-methoxyphenyl propyl azanium,gamma-amino-4-methoxy-benzenepropanol
2733725
3-amino-3-(4-methoxyphenyl)propan-1-ol
COC1=CC=C(C=C1)C(CCO)N
68208-24-2
C10H15NO2
181.235
MFCD01863268
WHWMCHUIUINGOD-UHFFFAOYSA-N
3-amino-3-4-methoxyphenyl propan-1-ol,3-amino-3-4-methoxyphenyl-1-propanol,3-amino-3-4-methoxy-phenyl-propan-1-ol,3-4-methoxyphenyl-dl-beta-alaninol,dl-beta-4-methoxyphenyl alaninol,3-amino-3-p-methoxyphenyl-1-propanol,benzenepropanol, gamma-amino-4-methoxy,1r-3-hydroxy-1-4-methoxyphenyl propyl azanium,gamma-amino-4-methoxy-benzenepropanol
2733725
3-amino-3-(4-methoxyphenyl)propan-1-ol
COC1=CC=C(C=C1)C(CCO)N
Quantity 1g
Color White
Infrared Spectrum Authentic
Melting Point 82°C to 86°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H4CH(NH)(CH)OH
Formula Weight 181.23
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Amino-3-(p-methoxyphenyl)-1-propanol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger