Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4,4-dimethylpentanoic acid hydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
336185-29-6
C7H15NO2·xH2O
145.2
MFCD03791052
AGUYRMXQCJEZJE-UHFFFAOYSA-N
3-amino-4,4-dimethylpentanoic acid xhydrate,3-amino-4,4-dimethylpentanoic acid hydrate,3-amino-4,4-dimethylpentanoic acid-water 1/1
53393206
3-amino-4,4-dimethylpentanoic acid;hydrate
CC(C)(C)C(CC(=O)O)N.O
336185-29-6
C7H15NO2·xH2O
145.2
MFCD03791052
AGUYRMXQCJEZJE-UHFFFAOYSA-N
3-amino-4,4-dimethylpentanoic acid xhydrate,3-amino-4,4-dimethylpentanoic acid hydrate,3-amino-4,4-dimethylpentanoic acid-water 1/1
53393206
3-amino-4,4-dimethylpentanoic acid;hydrate
CC(C)(C)C(CC(=O)O)N.O
Melting Point 223°C to 225°C
Infrared Spectrum Authentic
Quantity 500mg
Color White
Packaging Glass bottle
Water 14% max. (K.F.)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (H3C)3CCH(NH2)CH2COOH
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
Formula Weight 145.2
Physical Form Crystalline Flakes
Percent Purity 98%
Chemical Name or Material 3-Amino-4, 4-dimethylpentanoic acid hydrate, 98%
Show More Show Less