Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Amino-4-bromo-1-methyl-1H-pyrazole, 97+%, Alfa Aesar™
Click to view available options
Quantity:
0.25g
1g
5g
Specifications
Melting Point | 96°C to 100°C |
Quantity | 0.25g |
Formula Weight | 176.02 |
Percent Purity | ≥97% |
Chemical Name or Material | 3-Amino-4-bromo-1-methyl-1H-pyrazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures