Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-amino-4-chlorobenzenesulfonyl Fluoride 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
368-72-9
C6H5ClFNO2S
209.619
MFCD00007669
HNZYBSQZUOJCDN-UHFFFAOYSA-N
3-amino-4-chlorobenzenesulphonyl fluoride,benzenesulfonyl fluoride, 3-amino-4-chloro,3-amino-4-chlorophenyl fluorosulfone,3-amino-4-chlorophenylsulfonyl fluoride,benzenesulfonylfluoride, 3-amino-4-chloro,3-amino-4-chloro benzene sulphonyl fluoride,3-amino-4-chlorobenzene-1-sulfonyl fluoride,3-amino-4-chlorobenzenesulfonic acid fluoride,3-amino-4-chlorobenzene sulfonyl fluoride,3-amino-4-chloro-benzenesulfonic acid fluoride
97383
3-amino-4-chlorobenzenesulfonyl fluoride
C1=CC(=C(C=C1S(=O)(=O)F)N)Cl
368-72-9
C6H5ClFNO2S
209.619
MFCD00007669
HNZYBSQZUOJCDN-UHFFFAOYSA-N
3-amino-4-chlorobenzenesulphonyl fluoride,benzenesulfonyl fluoride, 3-amino-4-chloro,3-amino-4-chlorophenyl fluorosulfone,3-amino-4-chlorophenylsulfonyl fluoride,benzenesulfonylfluoride, 3-amino-4-chloro,3-amino-4-chloro benzene sulphonyl fluoride,3-amino-4-chlorobenzene-1-sulfonyl fluoride,3-amino-4-chlorobenzenesulfonic acid fluoride,3-amino-4-chlorobenzene sulfonyl fluoride,3-amino-4-chloro-benzenesulfonic acid fluoride
97383
3-amino-4-chlorobenzenesulfonyl fluoride
C1=CC(=C(C=C1S(=O)(=O)F)N)Cl
Infrared Spectrum Authentic
Quantity 5g
Color Brown to Brown-Yellow
Packaging Glass bottle
Melting Point 64°C to 68°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2NC6H3(Cl)SO2F
Formula Weight 209.62
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Amino-4-chlorobenzenesulfonyl fluoride
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger