Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-hydroxy-5-nitrobenzenesulfonic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
96-93-5
C6H6N2O6S
234.182
MFCD00035910
RHPXYZMDLOJTFF-UHFFFAOYSA-N
3844407
C1=C(C=C(C(=C1[N+](=O)[O-])O)N)S(=O)(=O)O
96-93-5
C6H6N2O6S
234.182
MFCD00035910
RHPXYZMDLOJTFF-UHFFFAOYSA-N
3844407
C1=C(C=C(C(=C1[N+](=O)[O-])O)N)S(=O)(=O)O
Color Undesignated
Quantity 5g
Formula Weight 234.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 3-Amino-4-hydroxy-5-nitrobenzenesulfonic Acid

EINECSNumber : (3)-2096

RTECSNumber : DB4900000

TSCA : No