Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-hydroxyphenylboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-4380040
Click to view available options
Quantity:
1 g
5 g
760990-10-1
C12H18BNO3
235.09
RXHWPAXQYXTPJB-UHFFFAOYSA-N
3-amino-4-hydroxyphenylboronic acid pinacol ester, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-amino-4-hydroxyphenylboronic acid, pinacol ester, 2-amino-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-amino-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
46738034
2-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)O)N
760990-10-1
C12H18BNO3
235.09
RXHWPAXQYXTPJB-UHFFFAOYSA-N
3-amino-4-hydroxyphenylboronic acid pinacol ester, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-amino-4-hydroxyphenylboronic acid, pinacol ester, 2-amino-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-amino-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
46738034
2-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)O)N
Melting Point 151°C to 152°C
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 235.09
Percent Purity 95%
Chemical Name or Material 3-Amino-4-hydroxyphenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit