Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-iodo-1H-pyrazole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
81542-51-0
C3H4IN3
208.99
CPVCVVMJPIOKMN-UHFFFAOYSA-N
4-iodo-1h-pyrazol-3-amine,3-amino-4-iodo-1h-pyrazole,4-iodo-2h-pyrazol-3-amine,4-iodo-3-aminopyrazole,3-amino-4-iodo-pyrazole,1h-pyrazol-5-amine, 4-iodo
12831626
4-iodo-1H-pyrazol-5-amine
C1=NNC(=C1I)N
81542-51-0
C3H4IN3
208.99
CPVCVVMJPIOKMN-UHFFFAOYSA-N
4-iodo-1h-pyrazol-3-amine,3-amino-4-iodo-1h-pyrazole,4-iodo-2h-pyrazol-3-amine,4-iodo-3-aminopyrazole,3-amino-4-iodo-pyrazole,1h-pyrazol-5-amine, 4-iodo
12831626
4-iodo-1H-pyrazol-5-amine
C1=NNC(=C1I)N
Melting Point 143°C to 146°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 208.99
Percent Purity 97%
Chemical Name or Material 3-Amino-4-iodo-1H-pyrazole

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning