Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-iodopyridine, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
105752-11-2
C5H5IN2
220.01
ZJRSKTXMSIVNAU-UHFFFAOYSA-N
3-amino-4-iodopyridine,4-iodo-pyridin-3-ylamine,3-pyridinamine, 4-iodo,4-iodo-3-pyridinamine,3-amine-4-iodopyridine,pubchem6681,4-iodo-3-pyridylamine,4-iodo-3-aminopyridine,4-iodo-3-pyridinylamine,4-iodopyridin-3-ylamine
819132
4-iodopyridin-3-amine
C1=CN=CC(=C1I)N
105752-11-2
C5H5IN2
220.01
ZJRSKTXMSIVNAU-UHFFFAOYSA-N
3-amino-4-iodopyridine,4-iodo-pyridin-3-ylamine,3-pyridinamine, 4-iodo,4-iodo-3-pyridinamine,3-amine-4-iodopyridine,pubchem6681,4-iodo-3-pyridylamine,4-iodo-3-aminopyridine,4-iodo-3-pyridinylamine,4-iodopyridin-3-ylamine
819132
4-iodopyridin-3-amine
C1=CN=CC(=C1I)N
Melting Point 65°C to 70°C
Quantity 5g
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 3-Amino-4-iodopyridine

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning