Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Amino-4-methoxybenzanilide, 98%

Click to view available options
Quantity:
10g
50g
120-35-4
C14H14N2O2
242.278
MFCD00017166
LHMQDVIHBXWNII-UHFFFAOYSA-N
3-amino-4-methoxybenzanilide,3-amino-p-anisanilide,benzamide, 3-amino-4-methoxy-n-phenyl,3-amino-4-methoxy benzanilide,benzanilide, 3-amino-4-methoxy,p-anisanilide, 3-amino,unii-v5y646aw8s,amba,3-amino-4-methoxy-n-phenyl-benzamide,3-amino-4-methoxyphenyl-n-benzamide
8426
3-amino-4-methoxy-N-phenylbenzamide
COC1=C(C=C(C=C1)C(=O)NC2=CC=CC=C2)N
120-35-4
C14H14N2O2
242.278
MFCD00017166
LHMQDVIHBXWNII-UHFFFAOYSA-N
3-amino-4-methoxybenzanilide,3-amino-p-anisanilide,benzamide, 3-amino-4-methoxy-n-phenyl,3-amino-4-methoxy benzanilide,benzanilide, 3-amino-4-methoxy,p-anisanilide, 3-amino,unii-v5y646aw8s,amba,3-amino-4-methoxy-n-phenyl-benzamide,3-amino-4-methoxyphenyl-n-benzamide
8426
3-amino-4-methoxy-N-phenylbenzamide
COC1=C(C=C(C=C1)C(=O)NC2=CC=CC=C2)N
Quantity 50g
Melting Point 154°C to 156°C
Formula Weight 242.28
Percent Purity 98%
Chemical Name or Material 3-Amino-4-methoxybenzanilide

EINECSNumber : 204-388-4

RTECSNumber : CV7400000

TSCA : Yes

Recommended Storage : Ambient temperatures