Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Amino-4-methylphenylboronic acid, 98%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
Specifications
Packaging | Glass Bottle |
Melting Point | 250.0°C |
Infrared Spectrum | Authentic |
Quantity | 1g |
Assay Percent Range | 97.5% min. (HPLC) |
Formula Weight | 150.98 |
Percent Purity | 98% |
Chemical Name or Material | 3-Amino-4-methylphenylboronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning