Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-methylpyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3430-27-1
C6H8N2
108.14
MFCD00128871
IBKMZYWDWWIWEL-UHFFFAOYSA-N
3-amino-4-methylpyridine,3-amino-4-picoline,3-pyridinamine, 4-methyl,4-methyl-3-pyridinamine,4-methyl-pyridin-3-ylamine,unii-9hn55rq07b,4-methyl-3-pyridylamine,4-methylpyridine-3-amine,3-amino-4-methyl pyridine,3-amino-4-methylpyridine 3-amino-4-picoline
137935
4-methylpyridin-3-amine
CC1=C(C=NC=C1)N
3430-27-1
C6H8N2
108.14
MFCD00128871
IBKMZYWDWWIWEL-UHFFFAOYSA-N
3-amino-4-methylpyridine,3-amino-4-picoline,3-pyridinamine, 4-methyl,4-methyl-3-pyridinamine,4-methyl-pyridin-3-ylamine,unii-9hn55rq07b,4-methyl-3-pyridylamine,4-methylpyridine-3-amine,3-amino-4-methyl pyridine,3-amino-4-methylpyridine 3-amino-4-picoline
137935
4-methylpyridin-3-amine
CC1=C(C=NC=C1)N
Packaging Glass bottle
Melting Point 101.0°C to 102.0°C
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material 3-Amino-4-methylpyridine

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye damage.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger